2-Acetyl-3-aminothiophene
Supplier: Thermo Scientific
GK00009.250
GK00009.1
MAYBGK00009.250EA
0
EUR
MAYBGK00009.250
MAYBGK00009.1
2-Acetyl-3-aminothiophene
2-Acetyl-3-aminothiophene
Formula:
C₆H₇NOS MW: 141.19 g/mol |
MDL Number:
MFCD00052585 CAS Number: 31968-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...