Sulphur trioxide pyridine complex, Sigma-Aldrich®
Supplier: Merck
84737-100G 84737-500G
SIAL84737-100GEA 86.4 EUR
SIAL84737-100G SIAL84737-500G
Sulphur trioxide pyridine complex, Sigma-Aldrich®
Sulphur trioxide pyridine complex
Formula: C₅H₅NO₃S
MW: 159.16 g/mol
MDL Number: MFCD00012437
CAS Number: 26412-87-3
EINECS: 247-683-3
UN: 3261
ADR: 8 II