(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: (R)-2-Isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, (2R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
H56077.AA H56077.XF
H56077.AAEA 596 EUR
H56077.AA H56077.XF
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine ≥98%
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
Formula: C₉H₁₆N₂O₂
MW: 184.24 g/mol
Density: 1.028
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040565
CAS Number: 109838-85-9