(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (R)-2-Isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, (2R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine
37286-1ML 37286-5ML
SIAL37286-1MLEA 148 EUR
SIAL37286-1ML SIAL37286-5ML
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, Sigma-Aldrich®
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
Formula: C₉H₁₆N₂O₂
MW: 184.24 g/mol
MDL Number: MFCD00040565
CAS Number: 109838-85-9