Diphenyleneiodonium chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

D2926-10MG D2926-50MG
SIALD2926-10MGEA 139 EUR
SIALD2926-10MG SIALD2926-50MG
Diphenyleneiodonium chloride, Sigma-Aldrich®
Diphenyleneiodonium chloride
Formula: C₁₂H₈ClI
MDL Number: MFCD00214165
CAS Number: 4673-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR