(Acetylmethylene)triphenylphosphorane, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(Triphenylphosphoranylidene)propan-2-one, 1-Triphenylphosphoranylidene-2-propanone

M-Clarity™ quality level = MQ200

158755-100G 158755-25G
SIAL158755-100GEA 579 EUR
SIAL158755-100G SIAL158755-25G
(Acetylmethylene)triphenylphosphorane, Sigma-Aldrich®
(Acetylmethylene)triphenylphosphorane
Formula: C₂₁H₁₉OP
MW: 318.36 g/mol
MDL Number: MFCD00008774
CAS Number: 1439-36-7
EINECS: 215-878-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR