PIG® small mobile container kit
Supplier: NEW PIG
KITE201 KITE301 KITE401 RFLE201 RFLE301 RFLE401
121-0061EA 223.25 EUR
121-0061 121-0067 121-0071 121-0117 121-0122 121-0125
PIG® small mobile container kit
Spill Control Materials
Fast, effective response to indoor and outdoor spills.
  • Durable, wheeled 120 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface
The weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).
Upozornení: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.