Multiple gel casters
Supplier: Hoefer
732-2525EA 194 EUR
HOEFSE618 HOEFSE209 HOEFSE614 HOEFSE218 HOEFSE613 HOEFSE678 HOEFSE238 HOEFSE616-2 HOEFSE612 HOEFSE237 HOEFSE215 HOEFSE213 HOEFSE201 HOEFSE233 HOEFSE231
Multiple gel casters
Multiple gel casters enable several gradient or homogeneous gels to be cast simultaneously.
  • Pour uniform gels from the top; pump gradient gels into an inlet port at the bottom of the unit
  • Sandwiches fill at the same time at the same rate from the same solution
  • Glass plate sandwiches seal leak free without the use of messy grease or tape
  • Once cast, gels can be wrapped in plastic and refrigerated for several weeks
Delivery information: Combs and spacers must be ordered separately.