2,4-Dichloro-5-methylpyrimidine 98%
Supplier: Thermo Fisher Scientific
Danger
398290050 398290010
ACRO398290050EA 196 EUR
ACRO398290050 ACRO398290010
2,4-Dichloro-5-methylpyrimidine 98%
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
Boiling Pt: 109 °C (11 mmHg)
Melting Pt: 26…28 °C
Density: 1.39 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance Light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %