3-(Difluoromethyl)pyridine ≥97%
Supplier: Thermo Scientific
Warning
H33189.03EA 123 EUR
H33189.03 H33189.MD
3-(Difluoromethyl)pyridine ≥97%
3-(Difluoromethyl)pyridine
Formula: C₆H₅F₂N
MW: 129,11 g/mol
Boiling Pt: 42 °C (6 mmHg)
Flash Pt: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD11226584
UN: 1993
ADR: 3,III