Indium 99,998 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
46661.G1
46661.G5
46661.G9
46661.G1EA
144
EUR
46661.G1
46661.G5
46661.G9
Indium 99,998 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Indium
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |