Palladium ≥99.95% (metals basis), wire, annealed, Ø 0.4 mm (0.0159 in)
Supplier: Thermo Fisher Scientific
46327.G1 46327.G5
46327.G1EA 578 EUR
46327.G1 46327.G5
Palladium ≥99.95% (metals basis), wire, annealed, Ø 0.4 mm (0.0159 in)
Palladium
Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6