L-Alanine
Supplier: G Bioscienes
Synonyms:
(S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
RC-013
RC-014
RC-013EA
177
EUR
RC-013
RC-014
L-Alanine
L-Alanine
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: 79 °C |
MDL Number:
MFCD00064410 CAS Number: 56-41-7 EINECS: 206-418-1 Merck Index: 13,00203 |