Mineral oil, white (petroleum), 20 cSt

Supplier: LGC Standards PROMOCHEM

PROOCON150-020-003EA 289 EUR
PROOCON150-020-003
Mineral oil, white (petroleum), 20 cSt
Mineral oil, white (petroleum)
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR