2-Methyl-1,3-benzoxazol-5-amine
Supplier: Apollo Scientific
Synonyms: 5-Amino-2-methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazol-5-amine
OR1878-1G OR1878-500MG
APOSOR1878-1GEA 180 EUR
APOSOR1878-1G APOSOR1878-500MG
2-Methyl-1,3-benzoxazol-5-amine
2-Methyl-1,3-benzoxazol-5-amine
Formula: C₈H₈N₂O
MW: 148.161 g/mol
MDL Number: MFCD00628970
CAS Number: 72745-76-7