Phenyl chloroacetate

Supplier: Thermo Scientific

KM03523.1 KM03523.10 KM03523.50
MAYBKM03523.1EA 0 EUR
MAYBKM03523.1 MAYBKM03523.10 MAYBKM03523.50
Phenyl chloroacetate
Phenyl chloroacetate
Formula: C₈H₇ClO₂
CAS Number: 620-73-5
EINECS: 210-650-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR