cis-11-Methyl-2-dodecenoic acid
Supplier: Cayman Chemical
Synonyms: (Z)-11-Methyldodec-2-enoic acid, DSF, Diffusible signal factor
10008123-1 10008123-10 10008123-5
CAYM10008123-1EA 108 EUR
CAYM10008123-1 CAYM10008123-10 CAYM10008123-5
cis-11-Methyl-2-dodecenoic acid
cis-11-Methyl-2-dodecenoic acid
Formula: C₁₃H₂₄O₂
MW: 212.33 g/mol
CAS Number: 677354-23-3
UN: 1231
ADR: 3 II