N-Isopropylcyclohexylamine 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: N-Isopropylcyclohexanamine
171760050 171761000 171760250
ACRO171760050EA 25.7 EUR
ACRO171760050 ACRO171761000 ACRO171760250
N-Isopropylcyclohexylamine 99%
N-Isopropylcyclohexylamine
Formula: C₉H₁₉N
MW: 141.2539 g/mol
Density: 0.85 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00003831
CAS Number: 1195-42-2
EINECS: 214-798-5
UN: 2734
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20°C, 589 nm)
Titration with HClO4 ≥98.5 %
HPLC ≥94 %