(4-Aminophenyl)acetonitrile 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide
A11304.06 A11304.14
A11304.06EA 51.1 EUR
A11304.06 A11304.14
(4-Aminophenyl)acetonitrile 99%
(4-Aminophenyl)acetonitrile
Purity: 99% CAS No.: 3544-25-0 Molecular Formula: C8H8N2 Formula Weight: 132.17 Flash Point: 110°(230°F) Melting Point: 42-46° Boiling Point: 312° Beilstein Reference: 1100108 UN No.: 3439 MDL No.: MFCD00007912
Formula: C₈H₈N₂
MW: 132.17 g/mol
Boiling Pt: 312 °C
Melting Pt: 42…46 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007912
CAS Number: 3544-25-0
EINECS: 222-587-4
UN: 3439
ADR: 6.1 III