4-Chloro-2-(trifluoromethyl)quinoline, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

681946-1G 681946-5G
SIAL681946-1GEA 55.4 EUR
SIAL681946-1G SIAL681946-5G
4-Chloro-2-(trifluoromethyl)quinoline, Sigma-Aldrich®
4-Chloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
MDL Number: MFCD00153105
CAS Number: 1701-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR