2-Hydroxy-1,4-naphthoquinone

Supplier: Molekula

MOLE30080734-100GEA 259 EUR
MOLE30080734-100G
2-Hydroxy-1,4-naphthoquinone
2-Hydroxy-1,4-naphthoquinone
Formula: C₁₀H₆O₃
MW: 174,16 g/mol
MDL Number: MFCD00001678
CAS Number: 83-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR