5-Hydroxy-1,4-naphthoquinone (juglone), Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Hydroxy-p-naphthoquinone, Juglone, 5-Hydroxy-1,4-naphthalenedione
M-Clarity™ quality level = MQ100
H47003-1G H47003-5G
SIALH47003-1GEA 66.4 EUR
SIALH47003-1G SIALH47003-5G
5-Hydroxy-1,4-naphthoquinone (juglone), Sigma-Aldrich®
5-Hydroxy-1,4-naphthoquinone (Juglone)
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
MDL Number: MFCD00001684
CAS Number: 481-39-0
EINECS: 207-567-5
UN: 2811
ADR: 6.1,III