(±)-3-Aminoquinuclidine dihydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

M-Clarity™ quality level = MQ100

100358-25G 100358-5G
SIAL100358-25GEA 235 EUR
SIAL100358-25G SIAL100358-5G
(±)-3-Aminoquinuclidine dihydrochloride, Sigma-Aldrich®
(±)-3-Aminoquinuclidine dihydrochloride
Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00137395
CAS Number: 6530-09-2
EINECS: 229-424-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR