Phenyl bromoacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate

404276-10G 404276-50G
SIAL404276-10GEA 124 EUR
SIAL404276-10G SIAL404276-50G
Phenyl bromoacetate, Sigma-Aldrich®
Phenyl bromoacetate
Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD00192391
CAS Number: 620-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR