Ethyl-4-bromocrotonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: (E)-Ethyl 4-bromobut-2-enoate, Ethyl trans-4-bromo-2-butenoate
M-Clarity™ quality level = MQ200
E13830-25G E13830-5G
SIALE13830-25GEA 120 EUR
SIALE13830-25G SIALE13830-5G
Ethyl-4-bromocrotonate, Sigma-Aldrich®
Ethyl 4-bromocrotonate
Formula: C₆H₉BrO₂
MW: 193.05 g/mol
MDL Number: MFCD00000247
CAS Number: 37746-78-4