(4-Acetamidophenyl)boronic acid pinacol ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide, 4-Acetylaminophenylboronic acid pinacol ester, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide

M-Clarity™ quality level = MQ100

518778-1G 518778-5G
SIAL518778-1GEA 45.6 EUR
SIAL518778-1G SIAL518778-5G
(4-Acetamidophenyl)boronic acid pinacol ester, Sigma-Aldrich®
(4-Acetamidophenyl)boronic acid pinacol ester
Formula: C₁₄H₂₀BNO₃
MW: 261.125 g/mol
MDL Number: MFCD02093722
CAS Number: 214360-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR