(2-Acetamidophenyl)boronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Acetamidobenzeneboronic acid

M-Clarity™ quality level = MQ100

683906-1G 683906-250MG
SIAL683906-1GEA 103 EUR
SIAL683906-1G SIAL683906-250MG
(2-Acetamidophenyl)boronic acid, Sigma-Aldrich®
(2-Acetamidophenyl)boronic acid
Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
MDL Number: MFCD02179474
CAS Number: 169760-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR