5-Chloro-2-phenoxyaniline, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Amino-4-chlorodiphenyl ether, 2-Amino-4-chlorophenyl phenyl ether, 5-Chloro-2-phenoxyaniline, 2-Amino-4-chloro-diphenyl ether

M-Clarity™ quality level = MQ100

562939-25G 562939-500G
SIAL562939-25GEA 87.5 EUR
SIAL562939-25G SIAL562939-500G
5-Chloro-2-phenoxyaniline, Sigma-Aldrich®
5-Chloro-2-phenoxyaniline
Formula: C₁₂H₁₀ClNO
MW: 219.6700 g/mol
MDL Number: MFCD00025217
CAS Number: 93-67-4
EINECS: 202-266-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR