trans-4-Chloro-β-nitrostyrene

Supplier: Thermo Scientific

Synonyms: 1-Chloro-4-(2-nitrovinyl)benzene

S03529.1 S03529.10 S03529.50
MAYBS03529.1EA 0 EUR
MAYBS03529.1 MAYBS03529.10 MAYBS03529.50
trans-4-Chloro-β-nitrostyrene
trans-4-Chloro-β-nitrostyrene
Formula: C₈H₆ClNO₂
MW: 183.5917 g/mol
MDL Number: MFCD00024825
CAS Number: 706-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR