β,3-Dinitrostyrene

Supplier: Thermo Scientific

Synonyms: 1-Nitro-3-(2-nitrovinyl)benzene

S13012.1 S13012.10
MAYBS13012.1EA 0 EUR
MAYBS13012.1 MAYBS13012.10
β,3-Dinitrostyrene
β,3-Dinitrostyrene
Formula: C₈H₆N₂O₄
MW: 194.15 g/mol
MDL Number: MFCD00052063
CAS Number: 882-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR