4-Acetamidobenzyl alcohol ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: N-[4-(Hydroxymethyl)phenyl]acetamide, 4-Acetamidobenzyl alcohol, N-(4-(Hydroxymethyl)phenyl)acetamide
H33454.03 H33454.06
H33454.03EA 59.3 EUR
H33454.03 H33454.06
4-Acetamidobenzyl alcohol ≥97%
4-Acetamidobenzyl alcohol
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016868
CAS Number: 16375-88-5