4-(2-(Dimethylamino)ethyl)phenol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hordenine

04476-100MG 04476-1G
SIAL04476-100MGEA 70.2 EUR
SIAL04476-100MG SIAL04476-1G
4-(2-(Dimethylamino)ethyl)phenol, Sigma-Aldrich®
4-(2-(Dimethylamino)ethyl)phenol
Formula: C₁₀H₁₅NO
MDL Number: MFCD00051462
CAS Number: 539-15-1
EINECS: 208-710-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR