α-(4-Pyridyl)benzhydryl alcohol

Supplier: Thermo Scientific

Synonyms: alpha,alpha-Diphenyl-4-pyridylmethanol

SPB06323.1 SPB06323.10 SPB06323.50
MAYBSPB06323.1EA 0 EUR
MAYBSPB06323.1 MAYBSPB06323.10 MAYBSPB06323.50
α-(4-Pyridyl)benzhydryl alcohol
α-(4-Pyridyl)benzhydryl alcohol
Formula: C₁₈H₁₅NO
MDL Number: MFCD00006434
CAS Number: 1620-30-0
EINECS: 216-584-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR