N-(3-Butynyl)phthalimide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-(But-3-yn-1-yl)isoindoline-1,3-dione
M-Clarity™ quality level = MQ100
630861-25G 630861-5G
SIAL630861-25GEA 252 EUR
SIAL630861-25G SIAL630861-5G
N-(3-Butynyl)phthalimide, Sigma-Aldrich®
N-(3-Butynyl)phthalimide
Formula: C₁₂H₉NO₂
MW: 199.2100 g/mol
MDL Number: MFCD03695464
CAS Number: 14396-90-8