General purpose gloves, ActivArmr® 52-502
Supplier: Ansell
52-547/10 52-547/9
ANSE52-547/10EA 101 EUR
ANSE52-547/10 ANSE52-547/9
General purpose gloves, ActivArmr® 52-502
Gloves
These cotton jersey gloves with nitrile coating effectively repel grease, oil and dirt and supersede the classic leather general purpose glove in medium-duty applications.
  • More flexible, more comfortable, far more hard-wearing than leather gloves
  • High performance nitrile impregnation offers superior resistance to abrasion and good protection against snags, cuts and puncture
  • Soft, flexible jersey lining offers a close, comfortable fit while safely cushioning the hand
  • Lining absorbs perspiration, keeping the hand dry and comfortable for longer
  • Excellent dry grip makes handling quicker and easier
  • Safely machine-washable: will not shrink, stiffen or lose its shape
Ideal for assembling metal parts and components, applications in gas and electricity supply, offshore applications, handling light castings, metal parts and cold components, injection moulding, mining and warehouse work.
Certifikace: EN 388:2016 (3111A); Cat. II