di-Lithium tetraborate
Supplier: Thermo Fisher Scientific
Synonyms: Lithium tetraborate, Dilithium tetraborate
96149.D9 96149.A1
96149.D9EA 0 EUR
96149.D9 96149.A1
di-Lithium tetraborate
di-Lithium tetraborate
Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524