3',4'-(Methylenedioxy)acetophenone 98%
Supplier: Thermo Fisher Scientific
Warning
330810050 330810250
ACRO330810050EA 38.7 EUR
ACRO330810050 ACRO330810250
3',4'-(Methylenedioxy)acetophenone 98%
3',4'-(Methylenedioxy)acetophenone
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 86…89 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005831
CAS Number: 3162-29-6
EINECS: 221-613-1
Order Now
Specification Test Results
Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 89 °C
GC ≥97.5 %