(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(2R)-(-)-Glycidyl p-toluenesulphonate, (2R)-(-)-Glycidyl tosylate
M-Clarity™ quality level = MQ200
540110-25G
540110-5G
SIAL540110-25GEA
67.6
EUR
SIAL540110-25G
SIAL540110-5G
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate, Sigma-Aldrich®
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate
Formula:
C₁₀H₁₂O₄S MW: 228.2649 g/mol |
MDL Number:
MFCD00010834 CAS Number: 113826-06-5 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...