Benzofuran-2-carboxylic acid 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Benzo[b]furan-2-carboxylic acid
233260010 233260050 233260250
ACRO233260010EA 34.1 EUR
ACRO233260010 ACRO233260050 ACRO233260250
Benzofuran-2-carboxylic acid 98%
Benzofuran-2-carboxylic acid
Formula: C₉H₆O₃
MW: 162.15 g/mol
Melting Pt: 195…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00005848
CAS Number: 496-41-3
EINECS: 207-818-9
Order Now
Specification Test Results
Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration with NaOH ≥97.5 %