3-Amino-2-(methylthio)benzotrifluoride

Supplier: Thermo Scientific

SEW03605.250 SEW03605.1
MAYBSEW03605.250EA 0 EUR
MAYBSEW03605.250 MAYBSEW03605.1
3-Amino-2-(methylthio)benzotrifluoride
3-Amino-2-(methylthio)benzotrifluoride
Formula: C₈H₈F₃NS
MDL Number: MFCD01764739
CAS Number: 256508-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR