1-(Trifluoromethyl)cyclopropanecarboxylic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

M-Clarity™ quality level = MQ200

680141-1G 680141-250MG
SIAL680141-1GEA 156 EUR
SIAL680141-1G SIAL680141-250MG
1-(Trifluoromethyl)cyclopropanecarboxylic acid, Sigma-Aldrich®
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Formula: C₅H₅F₃O₂
MDL Number: MFCD03093070
CAS Number: 277756-46-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR