Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Supplier: Thermo Scientific
Warning
Synonyms: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat
44031.03 44031.06
44031.03EA 741 EUR
44031.03 44031.06
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate
CAS No.: 35138-22-8 Molecular Formula: [Rh(C8H12)2]BF4 Formula Weight: 406.08 Physical Form: Powder MDL No.: MFCD00075045
Formula: C₁₆H₂₄BF₄Rh
MW: 406.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075045
CAS Number: 35138-22-8
EINECS: 460-220-1