Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
701629-1G
701629-250MG
SIAL701629-1GEA
503
EUR
SIAL701629-1G
SIAL701629-250MG
Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate, Sigma-Aldrich®
Bis(1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate
Formula:
C₁₆H₂₄F₆RhSb |
MDL Number:
MFCD11113031 CAS Number: 130296-28-5 |