(S)-(-)-2-o-Tolyl-CBS-oxazaborolidine 0.5 mol/l in toluene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

654302-25ML 654302-5ML
SIAL654302-25MLEA 252 EUR
SIAL654302-25ML SIAL654302-5ML
(S)-(-)-2-o-Tolyl-CBS-oxazaborolidine 0.5 mol/l in toluene, Sigma-Aldrich®
(S)-(-)-2-o-Tolyl-CBS-oxazaborolidine
Formula: C₂₄H₂₄BNO
MDL Number: MFCD07369782
CAS Number: 463941-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR