(S)-(-)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone, Sigma-Aldrich®

Supplier: Merck

Synonyms: (S)-tert-Butyl 2-(tert-butyl)-3-methyl-4-oxoimidazolidine-1-carboxylate

M-Clarity™ quality level = MQ100

337595-1G 337595-250MG
SIAL337595-1GEA 539 EUR
SIAL337595-1G SIAL337595-250MG
(S)-(-)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone, Sigma-Aldrich®
(S)-(-)-1-Boc-2-tert-butyl-3-methyl-4-imidazolidinone
Formula: C₁₃H₂₄N₂O₃
MW: 256.3400 g/mol
MDL Number: MFCD00075101
CAS Number: 119838-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR