Calcium sulphate, Drierite™ 6 mesh for drying
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
43057.P2 43057.TD 43057.XA
43057.P2EA 0 EUR
43057.P2 43057.TD 43057.XA
Calcium sulphate, Drierite™ 6 mesh for drying
Calcium sulphate
CAS No.: 7778-18-9 Physical Form: Granular, -6 Mesh Sensitivity: Hygroscopic Appearance: White Odour: Odourless Merck Reference: 14,1706 MDL No.: MFCD00010912
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
Density: 2.964 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 14,1706