3,4-Epoxy-1-butene, Sigma-Aldrich®
Supplier: Merck
Synonyms: Butadiene monoxide
MQ100
127574-1G 127574-5G
SIAL127574-1GEA 121 EUR
SIAL127574-1G SIAL127574-5G
3,4-Epoxy-1-butene, Sigma-Aldrich®
3,4-Epoxy-1-butene
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 65
Melting Pt: –135 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005149
CAS Number: 930-22-3
EINECS: 213-210-4
UN: 1993
ADR: 3 II