Scopoletin (7-hydroxy-6-methoxycumarin) 95%
Supplier: Thermo Scientific
Warning
Synonyms: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one
302900500 302900010 302902500
ACRO302900500EA 99.9 EUR
ACRO302900500 ACRO302900010 ACRO302902500
Scopoletin (7-hydroxy-6-methoxycumarin) 95%
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formula: C₁₀H₈O₄
MW: 192.1681 g/mol
Melting Pt: 200…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00006872
CAS Number: 92-61-5
EINECS: 202-171-9
Order Now
Specification Test Results
Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 200 to 207 °C
HPLC ≥94.0 %