Glycinamide hydrochloride (H-Gly-NH2.HCl), Sigma-Aldrich®
Supplier: Merck
Synonyms: H-Gly-NH2.HCl
M-Clarity™ quality level = MQ200
50070-10G 50070-50G
SIAL50070-10GEA 45 EUR
SIAL50070-10G SIAL50070-50G
Glycinamide hydrochloride (H-Gly-NH2.HCl), Sigma-Aldrich®
Glycinamide hydrochloride (H-Gly-NH2.HCl)
Formula: C₂H₇ClN₂O
MW: 110.54 g/mol
MDL Number: MFCD00013008
CAS Number: 1668-10-6
EINECS: 216-789-1