Cyclopentanecarbaldehyde ≥98% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cyclopentanecarboxaldehyde

Stabilization: Stabilised with hydroquinone (HQ)

H56609.03 H56609.06
H56609.03EA 51 EUR
H56609.03 H56609.06
Cyclopentanecarbaldehyde ≥98% stabilised
Cyclopentanecarbaldehyde

Stabilized with 25 ppm 1,4-benzoquinone

Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 140…141 °C
Density: 0.919
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060798
CAS Number: 872-53-7
EINECS: 212-829-7
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR